Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ (+/-)-2-Fluoromethamphetamine Hydrochloride Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco F0211ML
Description
A certified solution standard suitable for use as starting material in calibrators and controls for GC/MS or LC/MS methamphetamine testing methods in clinical toxicology, forensic analysis or urine drug testing. A fluoro analog of methamphetamine, 2-Fluoromethamphetamine is sold as a club/designer drug in the illicit market. The drug, also known as 2-FMA, produces stimulant and possibly empathogenic or psychoactive effects.Specifications
67-56-1 | |
MFCD29036939 | |
1.0 mg/mL in methanol (as free base) | |
Ampule | |
9.7°C (49.5°F) |
C10H14FN · HCl | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
203.68 | |
1 mL |
Safety and Handling
H331-H370
H311
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation