Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ 2-Mononitroglycerin Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco M0021ML
Description
2-Mononitroglycerin is a major plasma metabolite of the pharmaceutical vasodilator nitroglycerin. This Certified Spiking Solution™ is suitable for use in chromatography or mass spectrometry-based applications in pharmaceutical research or clinical toxicology. Nitroglycerin is used to treat heart conditions such as angina and chronic heart failure.Specifications
75-05-8, 620-12-2 | |
MFCD01075379 | |
100 μg/mL in acetonitrile | |
Ampule | |
1 mL |
C3H7NO5 | |
UN 1648 3 / PGII | |
137.09 | |
-20°C |
Safety and Handling
EINECSNumber : 200-835-2
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation