Learn More
CERILLIANT™ 25B-NBOMe-D3 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco B0621ML
Description
This stable-labeled internal standard is suitable for quantitation of 25B-NBOMe in LC/MS or GC/MS testing methods for forensic analysis, urine drug testing or clinical toxicology applications. Sold under street names such as New Nexus, N-Bomb, and 2CB, 25B-NBOMe is a very potent hallucinogen with active doses in the microgram range. This designer drug is a second generation 2C amine derivative of the phenethylamine 2C-B.Specifications
67-56-1 | |
UN 1230 - class 3 - PG 2 - Methanol | |
419.76 | |
-20°C |
C18H19D3BrNO3 · HCl | |
100 μg/mL in methanol (as free base) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation