Learn More
CERILLIANT™ 25I-NBOH Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco I0191ML
Description
25I-NBOH is a second generation 2C amine derived from attaching an NBOH (2-hydroxybenzyl) group to the amine group of 2C-I. 25I-NBOH is a hallucinogenic phenethylamine that appeared in the illicit drug market in late 2012. This certified solution standard is suitable for LC/MS or GC/MS 25I-NBOH testing applications in forensic analysis, urine drug testing or clinical toxicology.Specifications
67-56-1 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
449.71 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C17H20INO3 · HCl | |
1.0 mg/mL in methanol (as free base) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370-H412
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation