Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ 25I-NBOMe Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco I0161ML
Description
25I-NBOMe is a second generation 2C amine derived from attaching an NBOMe (2-methoxybenzyl) group to the amine group of 2C-I. Sold under street names such as N-bomb, Smiles and 25I, this designer drug produces vivid visual hallucinations similar to those of LSD. Suitable uses for this certified Snap-N-Spike™ solution include as starting material for calibrators or controls in 25I-NBOMe testing methods by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications.Specifications
67-56-1, 1043868-97-8 | |
MFCD18452514 | |
1.0 mg/mL in methanol (as free base) | |
Ampule |
C18H22INO3 · HCl | |
UN 1230 - class 3 - PG 2 - Methanol | |
463.74 | |
1 mL |
Safety and Handling
H331-H370
H311
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation