Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ 3,4-Dimethylmethcathinone Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco D0931ML
Description
3,4-Dimethylmethcathinone is a designer drug analog of the bath salt mephedrone. Also known as 3,4-DMMC, this stimulant drug is sold in many bath blends across Europe, Australia, and the US. This certified solution standard is suitable for use in bath salt testing methods by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications.Specifications
67-56-1, 1081772-06-6 | |
MFCD19303973 | |
1.0 mg/mL in methanol (as free base) | |
Ampule | |
1 mL |
C12H17NO · HCl | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
227.73 | |
-20°C | |
9.7°C (49.5°F) |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation