Learn More
CERILLIANT™ 3-Desmethylprodine Hydrochloride Solution, 1.0 mg/mL in Acetonitrile (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco D0901ML
Catalog No. 01-001-210
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
A certified Snap-N-Spike™ Solution suitable for LC/MS or GC/MS methods and applications in clinical toxicology, forensic analysis, urine drug testing, or pain prescription monitoring. MPPP or 3-Desmethylprodine is an opioid analgesic developed in the 1940s for use as a pharmaceutical but abused illegally today as a recreational drug. This designer drug is an analog of meperidine (Demerol™) but is listed by US DEA as a Schedule I drug since it has no medicinal use.
Specifications
Specifications
75-05-8, 4968-48-3
UN 1648 3 / PGII
283.79
-20°C
2°C (35.6°F)
C15H21NO2 · HCl
1.0 mg/mL in acetonitrile (as free base)
Ampule
1 mL
Safety and Handling
Safety and Handling
H332-H319
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation