Learn More
CERILLIANT™ 4-Chloro-2,5-Dimethoxyphenethylamine Hydrochloride Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco C1241ML
Description
2C-C, or 4-Chloro-2,5-Dimethoxyphenethylamine, is a psychedelic drug and phenethylamine of the 2C series. Sold under the street names 2C-C or 2CC the designer drug produces vivid visual hallucinations and unique among other 2C compounds, produces intense relaxing, almost sedating effects. This certified solution standard is suitable as a starting material for calibrators or controls in phenethylamine testing methods by GC/MS or LC/MS for applications in clinical toxicology, forensic analysis, or urine drug testing.Specifications
67-56-1, 88441-15-0 | |
UN 1230 - class 3 - PG 2 - Methanol | |
2C-C hydrochloride solution | |
Ampule | |
1 mL |
C10H14ClNO2 · HCl | |
1.0 mg/mL in methanol (as free base) | |
252.14 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation