Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ 4-Chloroisobutyryl fentanyl Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco C1850.5ML
Description
A certified Snap-N-Spike™ solution of 4-chloroisobutyryl fentanyl suitable for use as starting material in calibrators and controls for LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, or forensic analysis applications.Specifications
67-56-1 | |
MFCD30718352 | |
421.40 | |
0.5 mL |
C23H29ClN2O · HCl | |
100 μg/mL in methanol (as free base) | |
Ampule | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation