Learn More
CERILLIANT™ 4-Cl-alpha-PVP Hydrochloride Solution, 1 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco C1721ML
Description
4-chloro-α-Pyrrolidinovalerophenone, also known 4-Cl-α-PVP, is a derivative of the synthetic cathinone α-pyrrolidinovalerophenone. This illicit drug has stimulant properties and has been found in several bath salt blends across the US, Europe, and Australia. A certified Snap-N-Spike™ solution suitable for use in GC/MS or LC/MS testing methods for bath salt cathinones in clinical toxicology, urine drug testing or forensic analysis applications.Specifications
67-56-1, 5537-17-7 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
302.24 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C15H20ClNO · HCl | |
1 mg/mL in methanol (as free base) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation