Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ 4-Ethylmethcathinone Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco E0901ML
Description
Also known as 4-EMC, 4-Ethylmethcathione is often sold under the name 'NRG-2' as a mixture with other substituted cathinone derivatives. 4-EMC is a structural analog of 4-Methylethcathinone. This certified Snap-N-Spike™ solution is suitable as starting material in calibrators and controls for GC/MS or LC/MS synthetic cathinone testing methods in clinical toxicology, urine drug testing, or forensic analysis.Specifications
67-56-1 | |
MFCD27966861 | |
1.0 mg/mL in methanol (as free base) | |
Ampule | |
9.7°C (49.5°F) - closed cup |
C12H17NO · HCl | |
UN 1230 - class 3 - PG 2 - Methanol | |
227.73 | |
1 mL |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation