Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ 4-Fluoroamphetamine Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco F0181ML
Description
This Certified Spiking Solution™ is suitable for numerous applications in GC/MS or LC/MS testing of 4-fluoroamphetamine from clinical toxicology and urine drug testing to forensic analysis. A fluoro analog of amphetamine, 4-Fluoroamphetamine is sold in the illicit market as a club/designer drug under street names including R2D2, Flux and Flits. The drug produces stimulant and possibly empathogenic or psychoactive effects.Specifications
67-56-1, 64609-06-9 | |
MFCD01708102 | |
1.0 mg/mL in methanol (as free base) | |
Ampule | |
1 mL |
C9H12FN · HCl | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
189.66 | |
-20°C | |
9.7°C (49.5°F) |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation