Learn More
CERILLIANT™ 4-Fluoroisobutyryl fentanyl-13C6 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco F0580.5ML
Description
This stable-labeled internal standard is suitable for quantitation of 4-fluoroisobutyryl fentanyl levels in urine, serum, or plasma by LC/MS or GC/MS for clinical toxicology, forensic analysis, or isotope dilution methods. 4-Fluoroisobutyryl fentanyl is an opioid analgesic analog of fentanyl and has been sold as a designer drug on the illicit drug market.Specifications
67-56-1 | |
100 μg/mL in methanol | |
Ampule | |
0.5 mL |
C1713C6H29FN2O | |
374.44 | |
-20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation