Learn More
CERILLIANT™ 4-Hydroxyduloxetine acetate Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  MilliporeSigma Supelco H1171ML
Catalog No. 01-001-462
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
4-Hydroxyduloxetine is a major metabolite of duloxetine. Duloxetine, marketed in the US under the trade name Cymbalta™, is a serotonin-norepinephrine reuptake inhibitor (SNRI) used to treat major depressive disorder and to alleviate pain associated with diabetic neuropathy, fibromyalgia, and osteoarthritis. This Certified Spiking Solution™ is applicable for use in applications such as clinical toxicology, urine drug testing, and forensic analysis by LC-MS/MS or GC/MS.
Specifications
Specifications
67-56-1, 741693-83-4
100 μg/mL in methanol (as free base)
Ampule
C18H19NO2S · C2H4O2
373.47
1 mL
Safety and Handling
Safety and Handling
Recommended Storage : -70°C
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation