Learn More
CERILLIANT™ 4-Hydroxyduloxetine-beta-D-glucuronide Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco H1021ML
Description
4-Hydroxyduloxetine glucuronide is a metabolite of duloxetine. Duloxetine, marketed in the US under the trade name Cymbalta™ is a serotonin-norepinephrine reuptake inhibitor (SNRI) used to treat major depressive disorder and to alleviate pain associated with diabetic neuropathy, fibromyalgia, and osteoarthritis. This Certified Spiking Solution™ is applicable for use in applications such as clinical toxicology, urine drug testing, and forensic analysis by LC-MS/MS or GC/MS.Specifications
67-56-1, 741693-83-4 | |
MFCD09952222 | |
489.54 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C24H27NO8S | |
100 μg/mL in methanol | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation