Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ (+/-)-4-Methyl-N-ethyl-norephedrine Hydrochloride Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco M1591ML
Description
(±)-4-Methyl-N-ethyl-norephedrine HCl is a major urinary metabolite of 4-Methylethcathinone. 4-Methylethcathinone, also known as 4-MEC, is a synthetic cathinone found in numerous bath salt blends including NRG-1 and NRG-2. This certified Snap-N-Spike™ solution is suitable for LC/MS or GC/MS 4-MEC testing methods in forensic analysis, clinical toxicology, or urine drug testing.Specifications
67-56-1 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
229.75 | |
-20°C | |
9.7°C (49.5°F) |
C12H19NO · HCl | |
1.0 mg/mL in methanol (as free base) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation