Learn More
CERILLIANT™ 4-(Methylnitrosamine)-1-(3-pyridyl)-1-butanone-13C6 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco N0921ML
Description
Also known as nicotine-derived nitrosamine ketone, NNK is a potent carcinogen and biomarker of exposure to cigarette smoke. NNK, produced upon the curing of tobacco, is found in a variety of tobacco products including chewing tobacco, snuff, cigarettes, and cigars. This stable-labeled internal standard is suitable for use in LC-MS/MS or GC/MS applications in nicotine testing, environmental monitoring, or clinical and diagnostic testing of nicotine biomarkers.Specifications
67-56-1, 1346600-19-8 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
213.19 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C413C6H13N3O2 | |
100 μg/mL in methanol | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation