Learn More
CERILLIANT™ 5F-AMB Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco S1331ML
Description
5F-AMB, also known as 5-fluoro AMP and 5-fluoro MMB-PINACA, is a synthetic cannabinoid identified in illegal herbal products. This certified reference solution is suited for use in synthetic cannabinoid testing methods by LC/MS o GC/MS for applications in clinical toxicology, urine drug testing, and forensic analysis.Specifications
75-05-8, 1801552-03-3 | |
MFCD30718260 | |
363.43 | |
-20°C | |
6°C (42.8°F) - closed cup |
C19H26FN3O3 | |
100 μg/mL in acetonitrile | |
Ampule | |
1 mL |
Safety and Handling
H332-H319
H312
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation