Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ 6alpha/beta-Hydroxyoxymorphone-D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco H0981ML
Description
Also known as oxymorphol, 6α/β-Hydroxyoxymorphone is a major, active metabolite of the opiate oxymorphone. The metabolite is useful in cytochrome p450 studies and other pharmaceutical research. In addition to pharmaceutical research, this certified solution standard is also suitable for GC/MS or LC/MS applications in clinical toxicology, forensic analysis, pain management, and urine drug testing.Specifications
67-56-1 | |
MFCD16294876 | |
100 μg/mL in methanol | |
Ampule | |
1 mL |
C17H18NO4D3 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
306.37 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation