Learn More
CERILLIANT™ Acryl fentanyl-13C6 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco A1780.5ML
Description
This stable-labeled internal solution standard is suitable for quantitation of acryl fentanyl levels in urine, serum, or plasma by LC/MS or GC/MS for clinical toxicology, urine drug testing or forensic analysis applications. Acryl fentanyl, also known as acryloylfentanyl, is a highly potent opioid analgesic fentanyl analog that is sold as designer drug and abused recreationally.Specifications
67-56-1 | |
100 μg/mL in methanol (as free base) | |
Ampule | |
0.5 mL |
C1613C6H26N2O · HCl | |
376.87 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation