Learn More
CERILLIANT™ ADB-PINACA Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  MilliporeSigma Supelco S1281ML
Catalog No. 01-002-018
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Specifications
Specifications
67-56-1, 1801338-20-4
100 μg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C19H28N4O2
344.45
1 mL
Safety and Handling
Safety and Handling
H331-H370
H311
Recommended Storage : -20°C
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation