Learn More
CERILLIANT™ AH-7921-D3 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco A1141ML
Description
This stable-labeled internal standard is suitable for quantitation of AH-7921 levels in urine or blood by LC-MS or GC/MS methods in clinical toxicology, urine drug testing, or forensic analysis applications. AH-7921 is an opioid designer drug with 80% the potency of morphine when administered orally. First discovered in 2013, AH-7921 is an active ingredient of synthetic cannabis found in Japan.Specifications
67-56-1 | |
UN 1230 - class 3 - PG 2 - Methanol | |
368.74 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C16H19D3Cl2N2O · HCl | |
100 μg/mL in methanol (as free base) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation