Learn More
CERILLIANT™ alpha-Pyrrolidinopropiophenone Hydrochloride Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco P1001ML
Catalog No. 01-001-869
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Also known as α-PPP, this designer drug is a substituted cathinone found in bath salt blends and imitation 'ecstasy' tablets. α-PPP possesses stimulant effects and is similar in structure to the synthetic cathinones MDPV and pyrovalerone. This Certified Spiking Solution™ is suitable as starting material in calibrators and controls for synthetic cathinone testing methods by LC/MS or GC/MS in clinical toxicology, urine drug testing, or forensic analysis.
Specifications
Specifications
67-56-1, 92040-10-3
MFCD27966863
1.0 mg/mL in methanol (as free base)
Ampule
1 mL
C13H17NO · HCl
UN 1230 - class 3 - PG 2 - Methanol
239.74
-20°C
9.7°C (49.5°F) - closed cup
Safety and Handling
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation