Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ beta-Carotene(10,10′,11, 11′)-13C4 Solution, 100 μg/mL, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco V0591ML

Catalog No. 01-002-173


Add to Cart

Description

Description

This Snap-N-Shoot™ stable-labeled internal standard is suitable for quantification of β-carotene levels in serum or other matrices by LC/MS or GC/MS for clinical diagnostic, or food and nutrition testing applications. β-Carotene, also known as E160a when used as food coloring supplement, is a precursor to vitamin A. Serum levels of β-carotene are an adequate measurement of clinical status and can confirm the diagnosis of carotenoderma and is used to detect fat malabsorption. Low levels may be found in cases of steatorrhea.
Specifications

Specifications

109-99-9, 64-17-5
UN 1993 - class 3 - PG 2 - RQ - VP ≤ 110kPa - Flammable liquids, n.o.s., HI: all
540.84
1 mL
C3613C4H56
100 μg/mL (70:30 THF:ethanol with 0.1% BHT (w/v))
Ampule
-21.2°C (-6.2°F) - closed cup
Safety and Handling

Safety and Handling

Recommended Storage : -70°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

CERILLIANT is a registered trademark of Cerilliant Corporation