Learn More
CERILLIANT™ (+/-)-beta-Hydroxythiofentanyl-13C6 Hydrochloride Solution, 100 μg/mL, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco H1390.5ML
Description
An internal standard suitable for (+)-β-Hydroxythiofentanyl testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, urine drug testing, or forensic analysis. (+)-β-Hydroxythiofentanyl is an opioid analgesic and fentanyl analog. As of May 2016 (+)-β-Hydroxythiofentanyl is a Schedule I controlled substance in the United States.Specifications
67-56-1 | |
100 μg/mL in methanol (as free base) | |
Ampule |
C1413C6H26N2O2S · HCl | |
400.91 | |
0.5 mL |
Safety and Handling
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation