Learn More
CERILLIANT™ Camazepam Solution, 1 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco C1761ML
Description
Camazepam, marketed under the brand names Albego, Limpidon and Paxor, is a benzodiazepine psychoactive drug used for the short-term treatment of insomnia. This Certified Spiking Solution™ is suitable as starting material in calibrators and controls for GC/MS or LC/MS testing applications in forensic analysis, urine drug testing, clinical toxicology or therapeutic drug monitoring.Specifications
75-05-8, 36104-80-0 | |
UN 1648 3 / PGII | |
371.82 | |
-20°C | |
2°C (35.6°F) - closed cup |
C19H18ClN3O3 | |
1 mg/mL in acetonitrile | |
Ampule | |
1 mL |
Safety and Handling
H332-H319
H312
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation