Learn More
CERILLIANT™ Cannabicyclolic Acid (CBLA) Solution, 0.5 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco C1711ML
Description
Cannabicyclolic Acid (CBLA) is a non-psychoactive cannabinoid found in Cannabis. This Certified Spiking Solution™ is suitable for cannabicyclolic acid testing methods by GC/MS, LC/MS or HPLC for applications in testing of Cannabis potency or impurity profiling, pharmaceutical research, and forensic analysis. Cerilliant solution Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient, quantitative, US DEA-exempt solution format and with TK's for Canadian customers.Specifications
75-05-8, 40524-99-0 | |
UN 1648 3 / PGII | |
358.47 | |
-20°C | |
2°C (35.6°F) - closed cup |
C22H30O4 | |
0.5 mg/mL in acetonitrile | |
Ampule | |
1 mL |
Safety and Handling
H332-H319
H312
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation