Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Chenodeoxycholic acid-D4, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco C1471ML
Description
Chenodeoxycholic acid (CDCA) is a primary bile acid that facilitates the intestinal uptake of lipids and fat-soluble vitamins. Levels of chenodeoxycholic acid can serve as markers for inborn errors of bile acid synthesis and hepatobiliary disorders such as familial intrahepatic cholestasis and biliary atresia. This stable-labeled internal standard is suitable for quantitation of CDCA levels in LC/MS and GC/MS bile acid testing methods.Specifications
67-56-1, 99102-69-9 | |
MFCD00190400 | |
100 μg/mL in methanol | |
Ampule | |
1 mL |
C24H36D4O4 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
396.60 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation