Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ cis-4,7,10,13,16,19-Docosahexaenoic acid, 500 μg/mL in Ethanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco D1171ML
Description
An omega-3 fatty acid essential for normal brain growth and function, docosahexaenoic acid (DHA) plays an important role as a signaling factor in cells for both anti- and pro-inflammatory processes. Levels of docosahexaenoic acid as well as other fatty acids are analyzed by GC/MS or LC-MS/MS methods to monitor patients undergoing diet therapy for mitochondrial or peroxisomal disorders. This Certified Spiking Solution™ is suitable for use as starting material in the preparation of linearity standards, calibrators, or controls in mass spectrometry-based DHA testing applications such as assessment of cardiovascular disease risk and fatty acid defficiency, and detection and quantification of DHA in nutraceuticals and dietary supplements.Specifications
64-17-5, 6217-54-5 | |
MFCD00065722 | |
500 μg/mL in ethanol | |
328.49 | |
-20°C | |
14°C (57.2°F) - closed cup |
C22H32O2 | |
UN 1170 - class 3 - PG 2 - Ethanol, solution | |
DHA | |
Ampule | |
1 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation