Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ cis-5,8,11,14,17-Eicosapentaenoic acid, 500 μg/mL in Ethanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco E1131ML

Catalog No. 01-001-317


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

Eicosapentaenoic acid (cis-5,8,11,14,17), also known as EPA, is a long-chain omega-3 fatty acid. Studies suggests that EPA as well as other fatty acids can serve as biomarkers for cardiovascular disease, and nutritional and metabolic disorders. This certified solution standard is suitable for use as starting material in the preparation of linearity standards, calibrators, or controls in mass spectrometry-based EPA testing applications such as assessment of cardiovascular disease risk and fatty acid defficiency, and detection and quantification of EPA in nutraceuticals and dietary supplements.
Specifications

Specifications

64-17-5, 10417-94-4
CH3(CH2CH=CH)5(CH2)3CO2H
UN 1170 - class 3 - PG 2 - Ethanol
0.943 g/mL at 25°C (lit.)
302.45
n20/D 1.4977 (lit.)
14°C (57.2°F) - closed cup
-54 to 53°C (lit.)
MFCD00065716
500 μg/mL in ethanol
EPA, Timnodonic acid
Ampule
1 mL
Safety and Handling

Safety and Handling

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

CERILLIANT is a registered trademark of Cerilliant Corporation