Learn More
CERILLIANT™ Clobazam-8-chloro isomer-13C6 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco C1291ML
Description
A stable-labeled internal standard suited for quantification of clobazam levels in urine or other matrices by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis applications. Marketed under the brand names Frisium, Urbanol, and Onfi™, clobazam is a benzodiazepine used as an anxiolytic and anticonvulsant.Specifications
67-56-1 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
306.70 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
13C6C10H13ClN2O2 | |
100 μg/mL in methanol | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation