Learn More
CERILLIANT™ Cloniprazepam Solution, 1 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco C2051ML
Description
Cloniprazepam is a benzodiazepine derivative and prodrug for clonazepam, which is a potent, long-acting benzodiazepine with anxiolytic, anticonvulsant, muscle relaxant and hypnotic properties. This certified Snap-N-Spike™ solution is suitable for use in GC/MS or LC/MS applications in forensic analysis, clinical toxicology, urine drug testing, or pain prescription monitoring.Specifications
67-56-1 | |
MFCD30718343 | |
369.80 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C19H16ClN3O3 | |
1 mg/mL in methanol | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation