Learn More
CERILLIANT™ Cyanoguanidine (Metformin Related Compound A) Solution, 50 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco M1961ML
Catalog No. 01-001-669
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Cyanoguanidine, also known as Metformin Related Compound A, is an impurity produced during pharmaceutical manufacturing of metformin active pharmaceutical ingredients (API). Along with M-195-1ML Metformin Impurity Mixture, this solution-based Certified Reference Material is suited for use in metformin monograph testing methods by HPLC in testing applications ranging from API release testing to pharmaceutical research.
Specifications
Specifications
67-56-1, 461-58-5
MFCD00008066
50 μg/mL in methanol
Ampule
1 mL
C2H4N4
UN 1230 - class 3 - PG 2 - Methanol, solution
84.08
-20°C
9.7°C (49.5°F) - closed cup
Safety and Handling
Safety and Handling
H331-H370
H311
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation