Learn More
CERILLIANT™ Dabigatran-D3 Solution, 100 μg/mL (Acetonitrile with 10% 0.01 N HCl), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco D0991ML
Description
Dabigatran is an oral anticoagulant from the direct thrombin inhibitor class. Sold under the trade names Pradaxa™ and Prazaxa, dabigatran is used to prevent strokes or the formation of blood clots. This stable-labeled internal standard is suitable for quantification of dabigatran using LC/MS or LC-MS/MS methods for applications in clinical or diagnostic testing, clinical toxicology, or pharmaceutical research.Specifications
75-05-8, 1246817-44-6 | |
UN 1648 3 / PGII | |
474.53 | |
-20°C | |
2°C (35.6°F) |
C25H22D3N7O3 | |
100 μg/mL (acetonitrile with 10% 0.01 N HCl) | |
Ampule | |
1 mL |
Safety and Handling
H332-H319
H312
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation