Learn More
CERILLIANT™ delta9-Tetrahydrocannabinolic acid A-D3 (THCA-A-D3) Solution, 100 μg/mL, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco T1451ML
Specifications
75-05-8, 1548417-60-2 | |
100 μg/mL in acetonitrile | |
Ampule | |
1 mL |
C22H27D3O4 | |
361.49 | |
-70°C | |
6°C (42.8°F) - closed cup |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation