Learn More
CERILLIANT™ Deschloroketamine Hydrochloride Solution, 1 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco D1721ML
Description
Deschloroketamine, also known as DXE, DCK or 2′-Oxo-PCM, is a dissociative anesthetic sold as designer drug on the illicit drug market. This certified solution standard is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS testing applications from forensic analysis and clinical toxicology to urine drug testing.Specifications
67-56-1, 4631-27-0 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
239.74 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C13H17NO · HCl | |
1 mg/mL in methanol | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation