Learn More
CERILLIANT™ Desmethylene Paroxetine Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco D0841ML
Description
Desmethylene paroxetine is a major urinary metabolite of the SSRI antidepressant paroxetine. Paroxetine, marketed under trade names such as Paxil™ or Aropax, is used to treat many conditions in adults from major depression and obsessive-compulsive disorder to several anxiety disorders. Suitable uses for this certified solution standard include as a starting material for calibrators and controls in LC/MS or GC/MS paroxetine testing applications in clinical toxicology, urine drug testing or forensic analysis.Specifications
67-56-1, 1394861-12-1 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
353.82 | |
-70°C | |
11°C (51.8°F) |
C18H20FNO3 · HCl | |
1.0 mg/mL in methanol (as free base) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation