Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Desoxypipradrol Hydrochloride Solution, 1 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco D0821ML
Description
Desoxypipradrol, also referred to as 2-DPMP or 2-diphenylmethylpiperidine, is a stimulant and norepinephrine-dopamine reuptake inhibitor (NDRI). This designer drug, structurally similar to methylphenidate, is sometimes found in certain bath salt blends such as Ivory Wave. This Certified Spiking Solution™ is suitable as starting material in calibrators and controls for use in LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, or forensic analysis.Specifications
67-56-1, 5807-81-8 | |
MFCD09743875 | |
1 mg/mL in methanol | |
Ampule | |
1 mL |
C18H21N · HCl | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
287.83 | |
-20°C | |
9.7°C (49.5°F) |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation