Learn More
CERILLIANT™ Dexmethylphenidate Hydrochloride Solution, 1 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco D1701ML
Description
Dexmethylphenidate, sold under the brand name Focalin among others, is the active dextrorotatory enantiomer of methylphenidate. Dexmethylphenidate is a central nervous system (CNS) stimulant prescribed for treatment of attention deficit hyperactivity disorder (ADHD). This Certified Spiking Solution™; is applicable for use in LC/MS or GC/MS applications for urine drug testing, clinical toxicology and forensic analysis.Specifications
67-56-1 | |
1 mg/mL in methanol (as free base) | |
Ampule | |
9.7°C (49.5°F) - closed cup |
C14H19NO2 · HCl | |
269.77 | |
1 mL |
Safety and Handling
H331-H370
H311
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation