Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Dimethylone Hydrochloride Solution, 1 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco D1811ML
Description
A Certified Snap-N-Spike™ Solution for use in clinical toxicology, forensic analysis, or urine drug testing applications by LC/MS or GC/MS. Dimethylone, also known as bk-MDDMA, is a designer drug of the phenethylamine, amphetamine, and cathinone chemical classes with potential psychotropic effects.Specifications
67-56-1 | |
MFCD30718319 | |
257.71 | |
1 mL |
C12H15NO3 · HCl | |
1 mg/mL in methanol (as free base) | |
Ampule | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation