Learn More
CERILLIANT™ Diphenhydramine Impurity C Solution, 500 μg/mL, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco D1831ML
Description
Diphenhydramine is an active ingredient that is used to treat symptoms caused by histamines suffered because of the common cold or allergies. This certified solution standard contains Diphenhydramine Impurity C. This standard can be used for multiple HPLC applications including pharmaceutical testing, research, and Monograph testing methods.Specifications
75-05-8 | |
MFCD01727165 | |
370.71 | |
-20°C | |
6°C (42.8°F) - closed cup |
C17H20BrNO · HCl | |
500 μg/mL (acetonitrile:0.05% aqueous phosphoric acid (70:30) (v/v)) | |
Ampule | |
1 mL |
Safety and Handling
H332-H319
H312
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation