Learn More
CERILLIANT™ Diphenhydramine N-Oxide Hydrochloride Solution, 500 μg/mL, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco D1861ML
Description
Diphenhydramine N-oxide is a major metabolite of diphenhydramine. Diphenhydramine is an antihistamine sold over the counter under the trade name Benadryl and in other over the counter medications such as Tylenol PM, Advil PM, and Excedrin PM. Dihydramine is also consumed recreationally as a euphoriant and a deliriant.Specifications
75-05-8 | |
500 μg/mL (acetonitrile:0.05% aqueous phosphoric acid (70:30) (v/v)) | |
Ampule | |
1 mL |
C17H21NO2 · HCl | |
307.82 | |
-20°C | |
6°C (42.8°F) - closed cup |
Safety and Handling
H332-H319
H312
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation