Learn More
CERILLIANT™ Dothiepin HCl (cis/trans) Solution, 1 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco D1731ML
Description
Dothiepin, also known as Dosulepin and marketed as Prothiaden among others, is a tricyclic antidepressant (TCA) which is used in the treatment of depression. Dothiepin also has antihistamine, antiadrenergic, antiserotonergic, anticholinergic, and sodium channel-blocking effects. Due to its high toxicity and risk of overdose dothiepin is assayed in therapeutic drug monitoring to ensure patients remain within the drug's therapeutic range. This certified Snap-N-Spike™ solution is suitable for LC-MS/MS applications including therapeutic drug monitoring, clinical toxicology, urine drug testing or forensic analysis.Specifications
67-56-1, 897-15-4 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
331.90 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C19H21NS · HCl | |
1 mg/mL in methanol (as free base) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation