Learn More
CERILLIANT™ Estrogens CRM in Serum Solution, 1000 pg/mL (Sigmatrix Ultra), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco E5051ML
Description
Certified Reference Material of estrone and estradiol at indicated concentrations in serum for clinical research applications by mass spectrometry. Estrogens CRM in Serum is ready to be used as linearity standard or for calibration verification in LC-MS/MS estrogens testing methods. Separate options for individual levels of estrone and estradiol in serum at concentrations ranging from 10 to 1,000 pg/mL for each analyte, estrogens serum blank and 13C- or deuterium labeled internal standards of estrone and estradiol are also available.Specifications
50-28-2, 53-16-7 | |
1000 pg/mL (Sigmatrix Ultra) | |
Vial | |
1 mL |
C18H24O2 | |
272.38 | |
-20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation