Learn More
CERILLIANT™ Fluconazole Impurity Mixture Solution, 10 μg/mL (80:20 Water:Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco F0511ML
Description
Fluconazole belongs to the class of triazole antifungal antibiotic compounds showing solubility in water. Its mode of action involves disruption of the biosynthesis of ergosterol in fungal cells by attaching to the cytochrome P-450 dependent 14-α-sterol demethylase (P-450DM) enzyme. It is widely used in the treatment of a wide variety of infections caused by Candida sp. It is also used by the advanced HIV infection and AIDS patients in the management of oropharyngeal candidiasis.Specifications
75-05-8 | |
UN 1993C 3 / PGIII | |
288.28 | |
-20°C | |
24°C (75.2°F) |
C13H13FN6O | |
10 μg/mL (80:20 water:acetonitrile | |
Ampule | |
1 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation