Learn More
CERILLIANT™ FUB-APINACA Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco S1121ML
Catalog No. 01-002-008
Description
Description
FUB-APINACA, also known as FUB-AKB48, is an indazole-based synthetic cannabinoid that is sold on the illicit marked as designer drug. This certified solution standard is suitable for use in LC/MS or GC/MS synthetic cannabinoid testing applications including clinical toxicology, forensic analysis, and urine drug testing. For those synthetic cannabinoids currently controlled at the federal level, our certified solution standards of these substances are DEA-exempt, allowing laboratories to place orders without additional regulatory paperwork. For US customers, state specific regulatory requirements may apply.
Specifications
Specifications
67-56-1
UN 1230 - class 3 - PG 2 - Methanol, solution
403.49
-20°C
9.7°C (49.5°F) - closed cup
C25H26FN3O
100 μg/mL in methanol
Ampule
1 mL
Safety and Handling
Safety and Handling
H331-H370
H311
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation