Learn More
CERILLIANT™ Gabapentin Impurity Mixture Solution, 0.84-1.4 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco G0251ML
Description
Gabapentin is an antiepileptic drug, which is a structural analog of gamma-aminobutyric acid (GABA). It is used in the treatment of partial seizures in adults and children. This certified solution standard contains Gabapentin impurities including Impurity A, and B. This standard can be used for multiple HPLC applications including pharmaceutical testing, research, and Monograph testing methods.Specifications
67-56-1, 64744-50-9 | |
0.84 to 1.4 mg/mL in methanol | |
Ampule | |
1 mL |
C9H15NO | |
153.22 | |
-20°C | |
10°C (50°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation