Learn More
CERILLIANT™ Glutaryl-L-Carnitine-(N-Methyl-D3) Lithium Salt Solution, 100 μg/mL, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  MilliporeSigma Supelco A1811ML
Catalog No. 01-001-040
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
A stable-labeled internal standard of routinely tested Glutaryl-L-Carnitine at a concentration of 100 μg/mL. Acylcarnitines are biomarker for many inborn errors of metabolism that cause several metabolic diseases and are routinely tested in newborn screening programs. This stable-labeled internal standard is suitable for critical quantitative testing applications by LC/MS or GC/MS in research and clinical environments. The stable, quantitative solution format provides significant benefits to the user such as convenience of use, material savings, and increased efficiency of labor. Certified concentration value provides increased efficiency from less time remaking pools due to dilution errors or rerunning samples.
Specifications
Specifications
67-56-1
MFCD30830356
284.25
1 mL
C12D3H17NO6 · Li
100 μg/mL (90:10 methanol:1% aqueous formic acid)
Ampule
9.7°C (49.5°F) - closed cup
Safety and Handling
Safety and Handling
H331-H370
H311
Recommended Storage : -70°C
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation