Learn More
CERILLIANT™ Griseofulvin-D3 Solution, 1 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco G0241ML
Description
This stable-labeled internal standard is suitable for quantification of griseofulvin by LC/MS or GC/MS for therapeutic drug monitoring or clinical and diagnostic testing applications. Griseofulvin, sold under trade names such as Fulvicin, Grifulvin, Gris PEG and Grisactin, is an antifungal drug used used in the treatment and prevention of several types of dermatophytosis.Specifications
75-05-8 | |
UN 1648 3 / PGII | |
355.78 | |
-20°C | |
2°C (35.6°F) - closed cup |
C17H14D3O6Cl | |
1 mg/mL in acetonitrile | |
Ampule | |
1 mL |
Safety and Handling
H332-H319
H312
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation