Learn More
CERILLIANT™ Griseofulvin Solution, 2 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco G0231ML
Catalog No. 01-001-408
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Griseofulvin, sold under trade names such as Fulvicin, Grifulvin, Gris PEG and Grisactin, is an antifungal drug used used in the treatment and prevention of several types of dermatophytosis. This Certified Spiking Solution is suitable as starting material for calibrators, controls, or linearity standards for therapeutic drug monitoring or clinical and diagnostic testing of griseofulvin in patient blood, serum, or plasma samples by HPLC or LC-MS/MS.
Specifications
Specifications
75-05-8, 126-07-8
UN 1648 3 / PGII
352.77
-20°C
2°C (35.6°F) - closed cup
C17H17ClO6
2 mg/mL in acetonitrile
Ampule
1 mL
Safety and Handling
Safety and Handling
H332-H319
H312
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation